Learn More
Genscript Corporation CHO-K1/BB2 Stable Cell Line
Supplier:  Genscript Corporation M00182
Encompass
Recombinant CHO-K1 cells stably overexpress human bombesin receptor 2 (BB2) on the surface and contain high levels of G protein G q to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57080
Add to Cart
Request a Quote