Learn More
Genscript Corporation CHO-K1/CD16A 158F Stable Cell
Supplier:  Genscript Corporation M00586
Encompass
Recombinant CHO-K1 cells stably overexpress human CD16A and Fc epsilon receptor Ig (FcER1G) on the surface The surface expression of CD16A is validated by FACS analysis
Catalog No. 50-000-57298
Only null left
Add to Cart
Request a Quote