Learn More
Genscript Corporation CHO-K1/CD32C 13Gln Stable Cell
Supplier:  Genscript Corporation M00601
Encompass
Recombinant CHO-K1 cells stably overexpress Homo sapiens cell-type natural killer cells Fc gamma receptor IIc1 (Fc RIIc / CD32C) on the cell surface The surface expression of Fc RIIc is validated by FACS analysis This stable cell line is recommended for evaluating the binding affinity between Fc RIIc and Fc region of antibodies Fc fusion proteins or antibody-based biologics
Catalog No. 50-000-59221
Add to Cart
Request a Quote