Learn More
Genscript Corporation CHO-K1/G15/CCR1
Supplier:  Genscript Corporation M00522
Encompass
C-C chemokine receptor type 1 (CCR1) is a G protein-coupled receptor member of the CC chemokine subfamily of receptors Following interaction with its specific chemokine ligands like macrophage inflammatory protein 1 alpha (MIP-1 alpha) regulated on activation normal T expressed and secreted protein (RANTES) and monocyte chemoattractant protein 3 (MCP-3) it triggers the onset of a process known as chemotaxis GenScript s human CCR1-expressing stable subline is guaranteed to function properly in the calcium flux assay
Catalog No. 50-000-58735
Only null left
Add to Cart
Request a Quote