Learn More
Genscript Corporation CHO-K1/GHSR Stable Cell Line
Supplier:  Genscript Corporation M00189
Encompass
Recombinant CHO-K1 cells stably overexpress human growth hormone secretagogue receptor (GHSR) on the surface and contain high levels of G protein G q to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57388
Add to Cart
Request a Quote