Learn More
Genscript Corporation CHO-K1/GIP/G15 Stable Cell Li
Supplier:  Genscript Corporation M00486
Encompass
Recombinant CHO-K1 cells stably overexpress human gastric inhibitory polypeptide receptor (GIP) on the surface and contain high levels of G protein G s to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57446
Add to Cart
Request a Quote