Learn More
Genscript Corporation CHO-K1/HVEM Stable Cell Line
Supplier:  Genscript Corporation M00646
Encompass
This gene encodes a member of the TNF (tumor necrosis factor) receptor superfamily The encoded protein functions in signal transduction pathways that activate inflammatory and inhibitory T-cell immune response It binds herpes simplex virus (HSV) viral envelope glycoprotein D (gD) mediating its entry into cells Alternative splicing results in multiple transcript variants
Catalog No. 50-000-58086
Only null left
Add to Cart
Request a Quote