Learn More
Genscript Corporation CHO-K1/M2/G15 Stable Cell Lin
Supplier:  Genscript Corporation M00258
Encompass
Recombinant CHO-K1 cells stably overexpress human cholinergic receptor muscarinic 2 (M2) on the surface and contain high levels of G protein G i to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57861
Only null left
Add to Cart
Request a Quote