Learn More
Genscript Corporation CHO-K1/NK3 Stable Cell Line
Supplier:  Genscript Corporation M00201
Encompass
Recombinant CHO-K1 cells stably overexpress human tachykinin receptor 3 (NK3) on the surface and contain high levels of G protein G q to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57859
Only null left
Add to Cart
Request a Quote