Learn More
Genscript Corporation CHO-K1/NMU1 Stable Cell Line
Supplier:  Genscript Corporation M00191
Encompass
Neuromedin U receptor 1 (NMUR1) is a member of G-protein coupled receptor family A and is the most abundant in peripheral tissue particularly in small intestines and stomach This protein is a receptor for Neuromedin U (NMU-25) The first biological activity ascribed to NmU was smooth muscle contraction Neuromedin U has also been reported to increase arterial blood pressure and modify ion transport in the intestinal tract Finally NmU injected subcutaneously into rats has been reported to result in a short term increase in circulating ACTH levels and a long term increase in serum corticosterone levels suggesting a role in regulation of the hypothalamo-pituitary-adrenal axis
Catalog No. 50-000-59117
Only null left
Add to Cart
Request a Quote