Learn More
Genscript Corporation CHO-K1/OPRK1/G15 Stable Cell
Supplier:  Genscript Corporation M00290
Encompass
Recombinant CHO-K1 cells stably overexpress human opioid receptor kappa 1 (OPRK1) on the surface and contain high levels of G protein G i to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57738
Only null left
Add to Cart
Request a Quote