Learn More
Genscript Corporation CHO-K1/SST3/G15 Stable Cell L
Supplier:  Genscript Corporation M00285
Encompass
Recombinant CHO-K1 cells stably overexpress human somatostatin receptor 3 (SST3) on the surface and contain high levels of G protein G i to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57096
Add to Cart
Request a Quote