Learn More
Genscript Corporation CHO-K1/Tim3 Stable Cell Line
Supplier:  Genscript Corporation M00531
Encompass
Recombinant CHO-K1 cells stably overexpress human hepatitis A virus cellular receptor 2 (TIM3) on the surface The surface expression of TIM3 is validated by FACS analysis
Catalog No. 50-000-56707
Only null left
Add to Cart
Request a Quote