Learn More
Genscript Corporation CHO-K1/V1B Stable Cell Line
Supplier:  Genscript Corporation M00205
Encompass
The antidiuretic hormone arginine vasopressin (AVP) receptors are G protein-coupled receptors which consists of at least three types V1A (vascular/hepatic) and V1B (anterior pituitary) receptors which act through phosphatidylinositol hydrolysis to mobilize intracellular Ca sup 2 /sup and V2 (kidney) receptor which is coupled to adenylate cyclase V1B receptors are expressed in anterior pituitary where they mediate the release of ACTH Its peripheral actions such as antidiuresis contraction of vascular smooth muscle and stimulation of hepatic glycogenolysis are well characterized
Catalog No. 50-000-58623
Add to Cart
Request a Quote