Learn More
Genscript Corporation CHO-K1/V2/G15 Stable Cell Lin
Supplier:  Genscript Corporation M00170
Encompass
Arginine vasopressin (AVP) is a cyclic nonapeptide that acts by binding to a family of vasopressin receptors that includes V1a V1b and V2 receptors In particular V2 receptors are expressed in kidney where vasopressin exerts its antidiuretic action V1a and V1b couple to Gq and calcium release whereas V2 couples to Gs Mutations in V2 result in X-linked nephrogenic diabetes insipidus a syndrome in which the kidney is unable to concentrate urine leading to dehydration and hypernatremia Conversely elevated levels of AVP lead to hyponatremia in the syndrome of inappropriate antidiuretic hormone secretion (SIADH) congestive heart failure or cirrhosis and V2 selective antagonists have been developed to treat these conditions
Catalog No. 50-000-59044
Add to Cart
Request a Quote