Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIOCHEMPEG SCIENTIFIC INC CHO-PEG-CHO | 1g

Supplier:  BIOCHEMPEG SCIENTIFIC INC HO0040041K

Encompass_Preferred

CHO-PEG-CHO | Putity: ≥95% | MW: 1K | 1g

Catalog No. NC1711055


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.