Learn More
Eurofins Discoverx CHO-K1 BDKRB1 Gq Cell Line, 2 vials. (95-1011C2)
Supplier:  Eurofins Discoverx 951011C2
Encompass
The BDKRB1 Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. bradykinin) based activation of BDKRB1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses BDKRB1, a naturally Gq coupled GPCR. Upon activation of BDKRB1, the resulting calcium activity can be fluorescently assayed using the Calcium No WashPLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-4061
Add to Cart
Request a Quote