Learn More
Eurofins Discoverx PathHunter CHO-K1 CCR8 β-Arrestin Cell Line, 2 vials. (93-0196C2)
Supplier:  Eurofins Discoverx 930196C2
Encompass
The PathHunter CCR8 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CCL1) based activation of CCR8 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CCR8 and EA-tagged β-Arrestin-2. Activation of CCR8 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-199-9929
Add to Cart
Request a Quote