Learn More
Eurofins Discoverx PathHunter CHO-K1 EDNRA β-Arrestin Cell Line, 2 vials. (93-0257C2)
Supplier:  Eurofins Discoverx 930257C2
Encompass
The PathHunter EDNRA cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. endothelin-1) based activation of EDNRA GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged EDNRA and EA-tagged β-Arrestin-2. Activation of EDNRA stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0100
Add to Cart
Request a Quote