Learn More
Eurofins Discoverx cAMP Hunter CHO-K1 FSHR Gs Cell Line, 2 vials. (95-0119C2)
Supplier:  Eurofins Discoverx 950119C2
Encompass
The cAMP Hunter FSHR Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. FSH) based activation of FSHR GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type FSHR, a naturally Gs coupled GPCR. Upon activation of FSHR, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-3291
Only null left
Add to Cart
Request a Quote