Learn More
Eurofins Discoverx PathHunter CHO-K1 GALR2 β-Arrestin Cell Line, 2 vials. (93-0759C2)
Supplier:  Eurofins Discoverx 930759C2
Encompass
The PathHunter GALR2 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. galanin) based activation of GALR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GALR2 and EA-tagged β-Arrestin-2. Activation of GALR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1263
Only null left
Add to Cart
Request a Quote