Learn More
Eurofins Discoverx cAMP Hunter CHO-K1 GHRHR Gs Cell Line, 2 vials. (95-0076C2)
Supplier:  Eurofins Discoverx 950076C2
Encompass
The cAMP Hunter GHRHR Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. GRF) based activation of GHRHR GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type GHRHR, a naturally Gs coupled GPCR. Upon activation of GHRHR, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-2722
Add to Cart
Request a Quote