Learn More
Eurofins Discoverx cAMP Hunter CHO-K1 GPR109B Gi Cell Line, 2 vials. (95-0141C2)
Supplier:  Eurofins Discoverx 950141C2
Encompass
The cAMP Hunter GPR109B Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. 3-hydroxyoctanoic acid) based activation of GPR109B GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type GPR109B, a naturally Gi/Go coupled GPCR. Upon activation of GPR109B, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-3656
Add to Cart
Request a Quote