Learn More
Eurofins Discoverx PathHunter CHO-K1 mCCR5 β-Arrestin Cell Line, 2 vials. (93-0470C2)
Supplier:  Eurofins Discoverx 930470C2
Encompass
The PathHunter mCCR5 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CCL3/MIP-1α) based activation of mCCR5 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mCCR5 and EA-tagged β-Arrestin-2. Activation of mCCR5 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0598
Add to Cart
Request a Quote