Learn More
Eurofins Discoverx PathHunter CHO-K1 mCRHR1 β-Arrestin Cell Line, 2 vials. (93-0473C2)
Supplier:  Eurofins Discoverx 930473C2
Encompass
The PathHunter mCRHR1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CRH, sauvagine) based activation of mCRHR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mCRHR1 and EA-tagged β-Arrestin-2. Activation of mCRHR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0608
Add to Cart
Request a Quote