Learn More
Eurofins Discoverx PathHunter CHO-K1 mCRTH2 β-Arrestin Cell Line, 2 vials. (93-0584C2)
Supplier:  Eurofins Discoverx 930584C2
Encompass
The PathHunter mCRTH2 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin D2) based activation of mCRTH2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mCRTH2 and EA-tagged β-Arrestin-2. Activation of mCRTH2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0886
Only null left
Add to Cart
Request a Quote