Learn More
Eurofins Discoverx PathHunter CHO-K1 mPPYR1 (mNPY4R) β-Arrestin Cell Line, 2 vials. (93-0774C2)
Supplier:  Eurofins Discoverx 930774C2
Encompass
The PathHunter mPPYR1 (mNPY4R) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. pancreatic polypeptide) based activation of mPPYR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mPPYR1 and EA-tagged β-Arrestin-2. Activation of mPPYR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1287
Add to Cart
Request a Quote