Learn More
Eurofins Discoverx PathHunter CHO-K1 NPBWR1 β-Arrestin Cell Line, 2 vials. (93-0818C2)
Supplier:  Eurofins Discoverx 930818C2
Encompass
The PathHunter NPBWR1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. neuropeptides B/W) based activation of NPBWR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged NPBWR1 and EA-tagged β-Arrestin-2. Activation of NPBWR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1347
Add to Cart
Request a Quote