Learn More
Eurofins Discoverx PathHunter CHO-K1 rCRTH2 β-Arrestin Cell Line, 2 vials. (93-0580C2)
Supplier:  Eurofins Discoverx 930580C2
Encompass
The PathHunter rCRTH2 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin D2) based activation of rCRTH2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rCRTH2 and EA-tagged β-Arrestin-2. Activation of rCRTH2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0880
Only null left
Add to Cart
Request a Quote