Learn More
Eurofins Discoverx PathHunter CHO-K1 rGPR120 β-Arrestin Cell Line, 2 vials. (93-1031C2)
Supplier:  Eurofins Discoverx 931031C2
Encompass
The PathHunter rGPR120 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. free fatty acids) based activation of rGPR120 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rGPR120 and EA-tagged β-Arrestin-2. Activation of rGPR120 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1885
Add to Cart
Request a Quote