Learn More
Eurofins Discoverx PathHunter CHO-K1 SSTR4 β-Arrestin Cell Line, 2 vials. (93-0308C2)
Supplier:  Eurofins Discoverx 930308C2
Encompass
The PathHunter SSTR4 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. somatostatin) based activation of SSTR4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged SSTR4 and EA-tagged β-Arrestin-2. Activation of SSTR4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0229
Add to Cart
Request a Quote