Learn More
Eurofins Discoverx cAMP Hunter CHO-K1 TACR3 Gs Cell Line, 2 vials. (95-0092C2)
Supplier:  Eurofins Discoverx 950092C2
Encompass
The cAMP Hunter TACR3 Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. senktide, neurokinins) based activation of TACR3 GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type TACR3, a naturally Gs coupled GPCR. Upon activation of TACR3, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-2941
Add to Cart
Request a Quote