Learn More
Medchemexpress LLC Cholan-24-oic acid, 3,7,12-trihydroxy-, (3β,5β,7α,12α)- | 3338-16-7 | MFCD00272976 | 99.7% | 408.57 | C24H40O5 | 1 MG

Supplier: Medchemexpress LLC HYW5859221MG
3β-Cholic acid is a derivative of cholic acid found in human feces, provided as a white to off-white solid for laboratory research. It is used in studies of bile acid metabolism and related biochemical pathways and is supplied with analytical-grade purity appropriate for experimental applications.
- White to off-white solid, suitable for analytical use.
- High purity: 99.7% as supplied.
- Molecular formula C24H40O5; molecular weight 408.57.
- Soluble in DMSO (~12.5 mg/mL) with ultrasonic assistance.
- Powder stable at -20°C for up to 3 years.
- Intended for scientific research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.