Learn More
Medchemexpress LLC Cholecystokinin (26-33) free acid | 103974-46-5 | 99.8% | 1064.19 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP36515MG
Encompass_Preferred
Cholecystokinin (26-33) free acid, also known as CCK (26-33) free acid, is a cholecystokinin (CCK) fragment that can reduce food intake and gallbladder contraction. It is intended for research use only.
  • Cholecystokinin fragment
  • Reduces food intake
  • Reduces gallbladder contraction
  • For research use only
Catalog No. 50-003-98407