Learn More
CPC Scientific H-Asp-Tyr(SO3H)-Met-Gly-Trp-Met-Asp-Phe-NH2 (ammonium acetate salt) 10MG
SDP
Supplier:  CPC Scientific CCKS002D
Encompass
SEQUENCE: H-Asp-Tyr(SO3H)-Met-Gly-Trp-Met-Asp-Phe-NH2 (ammonium acetate salt)ONE-LETTER SEQUENCE: D-Tyr(SO3H)-MGWMDF-NH2MOLECULAR FORMULA: C49H62N10O16S3MOLECULAR WEIGHT: 1143.3STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [25126-32-3]SYNONYMS: Cholecystokinin (26-33), SQ 19,844, Sincalide, 5-L-methionine-3-10-caeruleinRESEARCH AREA: GastrointestinalREFERENCES:M.A. Ondetti et al., J. Amer. Chem. Soc., 92, 195 (1970)SKU(s): CCKS-002A, CCKS-002B, CCKS-002C, CCKS-002D
Catalog No. 50-210-9112