Learn More
Cayman Chemical Cholera Toxin (azide free), 1 mg
SDP
Supplier:  Cayman Chemical 196541
Encompass
A multimeric enterotoxin that catalyzes the ADP-ribosylation of the Gsα protein, which activates adenylate cyclase and increases the intracellular concentration of cAMP; causes the hypersecretion of chloride and bicarbonate ions, which results in increased fluid secretion and the severe diarrhea characteristic of the disease, cholera
Catalog No. NC1425699
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.