Learn More
Medchemexpress LLC Cholest-5-en-3-ol, 24,25-epoxy-, (3β,24S)- | 77058-74-3 | 98.1% | 400.64 | C27H44O2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW0401501MG
Encompass_Preferred
(24S,25)-Epoxycholesterol is an oxysterol that acts as an agonist of the liver X receptor (LXR). It is used in biochemical and cell biology research to study cholesterol metabolism, LXR-mediated signaling, and regulation of cholesterol homeostasis. The compound is supplied as a high-purity solid suitable for in vitro assays.
  • Used as an LXR agonist in biochemical and cell biology studies.
  • High purity for reproducible assay results.
  • Available in 1 mg, 5 mg, and 10 mg sizes for research use.
  • Suitable for in vitro assays investigating cholesterol metabolism and signaling.
Catalog No. 50-002-08905