Learn More
Medchemexpress LLC Cholest-5-en-3-yl [(2S)-2-amino-5-(diaminomethylideneamino)pentanoyl]glycinate, trifluoroacetate | 1609010-59-4 | >95.0% | 886.1 g·mol⁻¹ | C43H73F6N5O7 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1389135MG
Encompass_Preferred
2H-Cho-Arg (trifluoroacetate) is a steroid-based cationic lipid that couples a 2H-cholesterol skeleton to an L-arginine head group. It forms complexes with plasmid DNA and is used to facilitate gene transfection in cultured cells.
  • Steroid-based cationic lipid with a 2H-cholesterol skeleton and L-arginine head group.
  • Forms stable complexes with plasmid DNA, reducing DNA migration in electrophoretic assays.
  • Facilitates transfection of reporter genes in cell lines such as H1299.
  • Cellular uptake involves lipid raft and caveolae-mediated endocytosis pathways.
  • Supplied as the trifluoroacetate salt in solution; recommended storage -20 °C.
  • Reported purity greater than 95%.
Catalog No. 50-002-23418