Learn More
Medchemexpress LLC Cholestane-3,5-diol,6-[[2-(1H-imidazol-5-yl)ethyl]amino]-, (3β,5α,6β)- | 1191043-85-2 | 96.7% | 513.80 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1491325MG
Encompass_Preferred
Dendrogenin A (DDA) is a ligand for liver X receptor (LXR) that induces the expression of sodium/iodine symporter, and increases iodine uptake.
  • Induces expression of sodium/iodine symporter
  • Increases iodine uptake
  • Induces cell differentiation of MCF-7
  • Reactivates function of lactating cells
  • Induces expressions of TSH receptor, thyroid peroxidase, and thyroglobulin
  • Affects thyroid hormone generation
  • Exhibits cytotoxicity in cancer cell B-CPAP and 8505c
  • Arrests cell cycle at G0/G1 phase
Catalog No. 50-003-60852