Learn More
Medchemexpress LLC α-cholestane-d4 | 205529-74-4 | MFCD00216162 | 99.0% | C27H44D4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY132649S10MG
Encompass_Preferred
α-Cholestane-d4 is a deuterium-labeled analogue of α-cholestane used as a stable isotope internal standard and tracer in analytical workflows. It supports accurate quantitation, method development, and tracer studies in mass spectrometry and isotope-dilution assays, with high isotopic and chemical purity for reliable results.
  • Deuterium-labeled internal standard for mass spectrometry
  • High isotopic and chemical purity for accurate quantitation
  • Suitable for isotope-dilution assays and tracer studies
  • Available in small pack sizes for analytical use
  • Molecular formula C27H44D4 and CAS 205529-74-4 for identification
Catalog No. 50-002-59803