Learn More
Medchemexpress LLC Cholestenone-13C (4-cholesten-3-one-13C) | 68165-57-1 | 99.0% | 385.63 | C2613CH44O | 5 MG

Supplier: Medchemexpress LLC HY113365S15MG
Cholestenone-13C is a 13C-stable isotope labeled form of cholestenone intended for use as an analytical standard and metabolic tracer in biochemical research. It serves as an internal standard for quantitative mass spectrometry and for metabolic labeling studies, and is supplied in small research quantities with specified solubility and storage recommendations.
- Stable 13C-labeled cholestenone for tracer and standard applications.
- Chemical purity 99.0% for analytical reliability.
- Molecular weight 385.63 and formula C27H44O (13C labeled).
- Soluble in ethanol at 50 mg/mL; ultrasonic may be required.
- Recommended storage: powder -20°C for long term, 4°C for short term; in solvent -80°C for months.
- Available in small research sizes, including 5 mg vials.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.