Learn More
Cayman Chemical Cholesterol-3 4-13C2 10mg
SDP
Supplier:  Cayman Chemical 4027810 MG
Encompass_Preferred
An internal standard for the quantification of cholesterol by GC- or LC-MS
Catalog No. 50-294-2420