Learn More
Avanti Polar Lipids CHOLESTEROL 98% 1G
Supplier:  Avanti Polar Lipids 700000P1G
Encompass
Cholesterol, Cholesterol is Prepared from Wool Grease, Purity: > 98%, Molecular Weight: 386.66, Molecular Formula: C 27 H 46 O, Storage: -20 deg.C, Shelf Life: 12 Months, Powder form, White Color, Not Flammable, 1mg
Catalog No. NC9242129