Learn More
Medchemexpress LLC Cholesterol hemisuccinate Tris salt | 102601-49-0 | 99.98% | 607.86 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW010800A
Encompass_Preferred
Cholesterol hemisuccinate Tris salt, also known as Cholesterol hydrogen succinate Tris salt or CHS-Tris, is a compound exhibiting hepatoprotective and anticancer properties. It effectively inhibits Acetaminophen (AAP) hepatotoxicity, preventing AAP-induced hepatic apoptosis and necrosis. This compound also interferes with DNA polymerase and DNA topoisomerase, thereby hindering DNA replication, repair, and cell division, ultimately inhibiting tumor growth.
  • Can be used for buffer preparation
  • Amphiphilic molecule suitable for solubilization buffer preparation for glucan synthase assay
  • Can be used for immunoprecipitation buffer preparation
  • Functions as a membrane stabilizer
  • Can be used for drug delivery
Catalog No. 50-003-66411