Learn More
Medchemexpress LLC Cholesterol-PEG 600 | 69068-97-9 | <1% | 1055.60 g/mol | C68H110O8 | 250 MG

Supplier: Medchemexpress LLC HYW127558250MG
Cholesterol-PEG 600 is a cholesterol-polyethylene glycol conjugate supplied for research use, provided as an off-white to light yellow oil. It is designed to introduce cholesterol moieties into membranes or to increase water solubility of lipid-modified constructs for biochemical and formulation studies.
- Cholesterol-PEG conjugate for membrane anchoring and formulation.
- Soluble in water (~60 mg/mL) with ultrasonic and warming.
- Appearance: off-white to light yellow oil.
- Storage: store pure material at -20°C; in solvent, -80°C for long term; aliquot to avoid freeze-thaw.
- Certificate of analysis shows total cholesterol ~20.4% and free cholesterol <1%.
- Available in multiple pack sizes from milligram to gram quantities.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.