Learn More
Medchemexpress LLC Cholesterol-PEG-biotin | 99.9% | 2000 (average) | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW44071110MG
Encompass_Preferred
Cholesterol-PEG-Biotin (average MW 2000) is a biotin-conjugated, PEGylated cholesterol lipid used as a lipid anchor for liposome construction and surface biotinylation. The biotin moiety enables specific capture by avidin/streptavidin systems, while the PEG chain improves aqueous solubility and steric stabilization. Supplied as a powder for research use, it is intended for biochemical assays and liposome functionalization.
  • Biotin enables strong binding to avidin or streptavidin.
  • Pegylation (average MW 2000) enhances water solubility and steric stability.
  • Suitable as a lipid anchor for liposome surface functionalization.
  • Supplied as a powder with recommended storage at -20°C.
  • High reported purity (≈99.9%), suitable for biochemical assays.
Catalog No. 50-002-80497