Learn More
Medchemexpress LLC Cholesterol-PEG-maleimide | 95.1% | 2000 Da (average) | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW44071950MG
Encompass_Preferred
Cholesterol-PEG-maleimide (average MW 2000) is a cholesterol-terminated polyethylene glycol with a terminal maleimide group. It is a solid PEGylated lipid that self-assembles in water and is used to prepare liposomes, nanoparticles, and to conjugate thiol-containing molecules via stable thioether bonds.
  • Maleimide-functionalized PEG for thiol-specific conjugation.
  • Average molecular weight 2000 Da provides steric stabilization and PEGylation.
  • Solid powder form suitable for liposome and nanoparticle formulation.
  • Enables formation of stable thioether linkages with thiol-containing molecules.
  • Storage as powder at -20°C supports long-term stability.
Catalog No. 50-002-81639