Learn More
Medchemexpress LLC Cholesterol-SA-PEG-Azide | 90.0% | 2000 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1715625MG
Encompass_Preferred
Cholesterol-SA-PEG-Azide (MW 2000) is a lipid used for drug delivery. It consists of Cholesterol, Succinic acid, PEG units, and an azide group. The azide group allows it to undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing an alkyne group.
  • Contains cholesterol, succinic acid, PEG units, and an azide group.
  • Azide group enables copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc).
  • Can be used for drug delivery.
Catalog No. 50-003-86343