Learn More
Cayman Chemical Cholesteryl-d7 PalmItate 1mg
SDP
Supplier:  Cayman Chemical 90004221 MG
Encompass_Preferred
An internal standard for the quantification of cholesteryl palmitate by GC- or LC-MS
Catalog No. 50-289-4397