Learn More
Cayman Chemical Cholesteryl Erucate 50mg
SDP
Supplier:  Cayman Chemical 2646850 MG
Encompass_Preferred
A cholesterol ester; has been used as a standard for the quantification of cholesterol esters in human meibomian gland secretions
Catalog No. 50-293-4487